Anti-MALT1 Mouse Monoclonal Antibody [clone: MT1/410] (Alexa Fluor 647)
Leverandør: Novus Biologicals
NBP2-33054AF64
NOVUNBP2-33054AF64EA 8225 NOK
NOVUNBP2-33054AF64
Anti-MALT1 Mouse Monoclonal Antibody [clone: MT1/410] (Alexa Fluor 647)
Antistoffer
Mucosa-associated lymphoid tissue lymphoma translocation protein 1 (MALT1), also known as MLT1 or paracaspase 1, is a member of the human paracaspase family and an interacting partner of B-cell lymphoma 10 (Bcl-10) (1). In vitro, In vitro, MALT1 synergises with BCL10 to enhance nuclear factor B activation (2) and to mediate IB kinase (IKK) activation by facilitating the ubiquitinylation of the NF-B essential modulator (NEMO) (3).
MALT1 is found in recurrent chromosomal translocations in non-Hodgkin B-cell lymphoma of the mucosa-associated lymphoid tissue (MALT). One common translocationMore Product Information