Anti-PKC theta Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NBP2-55717
NOVUNBP2-55717EA 7450 NOK
NOVUNBP2-55717
Anti-PKC theta Rabbit Polyclonal Antibody
Antistoffer
Type:
Antigen: PKC theta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR