Goat Anti-Rat IgG2a Secondary Antibody

Leverandør: Novus Biologicals
NBP2-68486
NOVUNBP2-68486EA 6650 NOK
NOVUNBP2-68486
Goat Anti-Rat IgG2a Secondary Antibody
Antistoffer
Goat anti Rat IgG2a antibody recognises rat IgG2a as assessed by immunoelectrophoresis and ELISA. The antibody may cross-react with IgG2a from other species.

Recommended Dilutions: ELISA: 1 to 10 µg/ml, Flow Cytometry, Western Blot
Preservative: 0.1% Sodium Azide

Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn IgG2a
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Rat
Vert Goat
Isotype IgG
Western blot Yes
ELISA Yes
Flowcytometri Yes
Konsentrasjon 1 mg/ml
Kyssabsorpsjon Yes
Lagringsbuffer PBS
Lagringstemp Store at 4 °C
Renhet Affinity purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR