Anti-MALT1 Mouse Monoclonal Antibody [clone: MT1/410] (Alexa Fluor 488)

Leverandør: Novus Biologicals
NBP233054AF488
NOVUNBP233054AF488EA 8225 NOK
NOVUNBP233054AF488
Anti-MALT1 Mouse Monoclonal Antibody [clone: MT1/410] (Alexa Fluor 488)
Antistoffer
MALT1 Monoclonal specifically detects MALT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: MALT1
Clonality: Monoclonal
Clone: MT1/410
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR