Anti-DHX37 Rabbit Polyclonal Antibody

Leverandør: Novus Biologicals
NBP3-18056
NOVUNBP3-18056EA 5900 NOK
NOVUNBP3-18056
Anti-DHX37 Rabbit Polyclonal Antibody
Antistoffer
DHX37 is a DEAD box protein. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases.

They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division.This gene encodes a DEAD box protein. DEAD box proteins, characterised by the conservedMore Product Information
Bestill nå

SPESIFIKASJONER

Antigen-navn DHX37
Environmentally Preferable
Antigen synonymer probable ATP-dependent RNA helicase DHX37|MGC2695|MGC4322|DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 37|DEAH (Asp-Glu-Ala-His) box polypeptide 37|KIAA1517MGC46245|DDX37|DEAH box protein 37|EC 3.6.4.13|FLJ41974|EC 3.6.1|DEAD/DEAH box helicase DDX37
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human,Mouse
Vert Rabbit
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Immunogen Synthetic peptide made to an internal portion of human DHX37 (between amino acids 650-700)
Konsentrasjon 1,0 mg/ml
Lagringsbuffer PBS, contains 0,02% sodium azide
Lagringstemp Store at 4 °C short term, aliquot and store at −20 °C long term, avoid freeze-thaw cycles
UniProtKB Q8IY370
Renhet Immunogen affinity purified
Uttestet applikasjon Immunocytochemistry/ Immunofluorescence 1 - 2 µg/ml
Immunohistochemistry 1:200
Immunohistochemistry-Paraffin 1:200
Western Blot 1 - 2 µg/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR