Anti-PRKCQ Rabbit Monoclonal Antibody [clone: HL1157]
Leverandør: Novus Biologicals
NBP3-25607
NOVUNBP3-25607EA 8300 NOK
NOVUNBP3-25607
Anti-PRKCQ Rabbit Monoclonal Antibody [clone: HL1157]
Antistoffer
Members of the protein kinase C (PKC) family play a key regulatory role in variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function. PKCs were originally identified as serine/threonine protein kinases whose activity was dependent on calcium and phospholipids. Diacylglycerols (DAG) and tumor promoting phorbol esters bind to and activate PKC.
PKCs can be subdivided into at least two major classes including conventional (c) PKC isoforms (alpha, beta I, beta II and gamma) and novel (n) PKC isoforms (delta, episilon, zeta,More Product Information