Center Well dish for IVF, Nunc™
Leverandør: Thermo Fisher Scientific
150260
NUNC150260EA 0 NOK
NUNC150260
Center Well dish for IVF, Nunc™
Dyrkningsplater Cellekulturskåler
Non treated surface, sterile (SAL 10⁻⁶), with lid. These IVF centre well dishes meet the exacting requirements of IVF testing, certification, and validation, providing consistently reliable, high quality results during embryo and oocyte washing.
  • Perfect angle of reservoir well walls for locating and easy washing of samples
  • True flat bottom for maintaining optimal temperature
  • Easy grip design for increased safety
  • Certificates of conformity available for every lot
Produced under ISO 13485 with full lot traceability
Sertifiseringer: Produced under ISOMore Product Information