Supelco® Organic Reference Standard, F.A.M.E. Mix RM-3
Leverandør: Merck
O7256-1AMP
O7256-1AMPEA 658 NOK
O7256-1AMP
Supelco® Organic Reference Standard, F.A.M.E. Mix RM-3
Standarder Organiske standarder
Myristic acid methyl ester (C14:0) 1,0% Palmitic acid methyl ester (C16:0) 4,0% Stearic acid methyl ester (C18:0) 3,0% Oleic acid methyl ester (C18:1cis-9) 45,0% Linoleic acid methyl ester (C18:2cis-912) 15,0% Linolenic acid methyl ester (C18:3cis-91215) 3,0% Arachidic acid methyl ester (C20:0) 3,0% Behenic acid methyl ester (C22:0) 3,0% Erucic acid methyl ester (C22:1cis-13) 20,0% Lignoceric acid methyl ester (C24:0) 3,0%.
  • GHS signal: GHS05