Supelco® Organic Reference Standard, F.A.M.E. Mix RM-6

Leverandør: Merck

O7631-1AMP
O7631-1AMPEA 658 NOK
O7631-1AMP
Supelco® Organic Reference Standard, F.A.M.E. Mix RM-6
Standarder Organiske standarder

Myristic acid methyl ester (C14:0) 2,0% Palmitic acid methyl ester (C16:0) 30,0% Palmitoleic acid methyl ester (C16:1cis-9) 3,0% Stearic acid methyl ester (C18:0) 14,0% Oleic acid methyl ester (C18:1cis-9) 41,0% Linoleic acid methyl ester (C18:2cis-912) 7,0% Linolenic acid methyl ester (C18:3cis-91215) 3,0%.


  • GHS signal: GHS05

Bestill nå

SPESIFIKASJONER

Synonym(s) AOCS Reference Mixtures; AOCS No. 6 (RM-6); Oil Reference Standard, AOCS No. 6; Oil Reference Standard, AOCS
Environmentally Preferable
M-Clarity quality level MQ100
Fraktforhold DICE
Lagringsforhold –20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR