Light duty microplate shaker

Leverandør: OHAUS
30391907
OHAU30391907EA 14175 NOK
OHAU30391907
Light duty microplate shaker
Risteapparater Risteapparater, mikroplater
This light duty microplate shaker is ideal for immunoassays and general microplate shaking, and is designed to be used in a variety of environmental conditions. Ideal for ELISA assays and DNA studies.

  • Takes up less space and easily fits into most hoods and incubators
  • Variable speed microprocessor control provides consistent uniform shaking action and will display last set-point and will restart if power is interrupted
  • Speed ramping feature slowly increases speed to desired set-point to avoid splashing
  • Spill-resistant design ensures fluids are channelled
More Product Information
Bestill nå

SPESIFIKASJONER

Modell SHLDMP03DG
Environmentally Preferable
Speed 100 - 1200 min⁻¹
Hastighetsnøyaktighet ±2 %
kretsløp 3 mm
Max. load 4 microplates or 2 micro-tube racks
Effekt 25 W
Ristebevegelse Orbital
Weight 10,9 kg
W×D×H 413×254×102 mm
Dimensjon skål 279×197 mm
Ur 1 s – 160 h
Elektronisk 230 V, 5,0 A, 50/60 Hz

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR