Incubating orbital microplate shakers, light duty
Leverandør: OHAUS
30391926 30391933
OHAU30391926EA 30650 NOK
OHAU30391926 OHAU30391933
Incubating orbital microplate shakers, light duty
Risteapparater Risteinkubatorer
These light duty incubating shakers are designed to incubate samples from 10 °C below ambient to 65 °C depending on the model. The units are ideal for shaking four microplates, deep-well plates, or two micro-tube racks.
  • Variable speed microprocessor control provides consistent uniform shaking action and will display last set-point and will restart if power is interrupted
  • Audible and visual signals will activate when system detects an obstruction or tray overload
  • Caution hot indicator symbol illuminates when the temperature of the air in the chamber reaches