Waving shaker, digital
Leverandør: OHAUS
30391949
OHAU30391949EA 15550 NOK
OHAU30391949
Waving shaker, digital
Risteapparater Risteapparater, roterende/gyngende
This digital waving shaker provides precise speed control and a smooth, low foaming, three dimensional, 'wave' motion. Ideal for use in a wide range of laboratory applications and designed to be used in a variety of environmental conditions. Tray includes a non-skid rubber mat that is suitable for holding Petri dishes.
  • Takes up less space and easily fits into most hoods and incubators
  • Aluminum base offers durability and added stability
  • Audible and visual signals will activate when system detects an obstruction or overload of the tray
  • Speed ramping feature