Incubating waving shaker
Leverandør: OHAUS
30391982
OHAU30391982EA 43800 NOK
OHAU30391982
Incubating waving shaker
Risteapparater Risteinkubatorer
This incubating waving shaker combines vertical and horizontal 'wave' motion with general purpose incubation in one compact bench top unit. Polycarbonate lid allows samples to be viewed without disturbing internal temperature.
  • LED displays for temperature, speed and tilt angle and time
  • Slowly increases speed to desired set-point to avoid splashing
  • In timed mode, alarm will sound when the time reaches zero
  • Spill-resistant design ensures fluids are channelled away from internal components
  • Speed ramping feature slowly increases speed to desired set-point to