Vortex mixer, digital
Leverandør: OHAUS
30392136
OHAU30392136EA 13025 NOK
OHAU30392136
Vortex mixer, digital
Risteapparater Reagensrørrister
This heavy-duty digital vortex mixer is ideal for applications that demand repeatable results. Mixer features touchpad controls and LED displays for accurate speed and time (minutes and seconds) results. The heavy-duty design and efficient motor allow this mixer to operate in continuous duty mode and handle all accessories over the entire speed range.
  • Variable speed microprocessor control provides consistent uniform mixing action
  • Timer will display elapsed time or, when programmed to user defined time limits, the unit will shut off when time reaches zero