Vortex mixer, analogue
Leverandør: OHAUS
30392141
OHAU30392141EA 10750 NOK
OHAU30392141
Vortex mixer, analogue
Risteapparater Reagensrørrister
This heavy-duty analogue vortex mixer is a variable speed analogue mixer that is designed for continuous duty. The heavy-duty design and efficient motor allow this mixer to operate in continuous duty mode and handle all accessories over the entire speed range.
  • Economical alternative to digital models
  • Can be used at 4 to 40 °C (from 20 to max. 85% relative humidity, non condensing)
Two modes of operation: 'Touch' mode for mixing tubes when cup head or universal holder with cover is depressed, or 'On' mode when using any of the accessory attachments forMore Product Information