Guardian 7000 magnetic hotplate stirrers
Leverandør: OHAUS
30500603 30500613 30500623
OHAU30500603EA 13275 NOK
OHAU30500603 OHAU30500613 OHAU30500623
Guardian 7000 magnetic hotplate stirrers
Omrørere Magnetrørere med varmeplate
Durable magnetic hotplate stirrers with superior safety features and intelligent performance.
  • SmartPresence™ and SmartLink™ technologies allow unsupervised operation
  • SafetyHeat™ system protects the laboratory
  • Powerful, consistent stirring performance
  • Customised performance for precise applications
OHAUS Guardian 7000 magnetic hotplate stirrers are designed with outstanding safety features and superior heating and mixing performance. The SmartHousing™ is chemical resistant with an innovative, easy-to-clean design that channels spills away from internalMore Product Information