Trinocular Metallurgical Microscope, 500X, B-510MET
Leverandør: OPTIKA
B-510MET
OPTKB-510METEA 55700 NOK
OPTKB-510MET
Trinocular Metallurgical Microscope, 500X, B-510MET
Mikroskop
Advanced routine laboratory microscope with IOS W-PLAN MET objectives and metallurgical attachment with the exclusive X-LED3 lighting source for incident illumination only. The NCG (no cover glass) objectives are especially designed for microscopy use without a cover slip ideal for metallographic samples and other opaque specimens.
Objectives (strain-free):
IOS W-PLAN MET 5X/0,12
IOS W-PLAN MET 10X/0,25
IOS W-PLAN MET 20X/0,40
IOS W-PLAN MET 50X/0,75
All with anti-fungus treatment