HBO Fluorescence Microscope, IM-300
Leverandør: OPTIKA
IM-300FL4
OPTKIM-300FL4EA 0 NOK
OPTKIM-300FL4
HBO Fluorescence Microscope, IM-300
Mikroskop
Advanced inverted microscope for brightfield and fluorescence observations with Semi-Apo IOS LWD U-PLAN F objectives to enhance the visibility of the sample and increase the overall contrast.
The HBO fluorescence illuminator provides an outstanding flexibility of use, standing the blue and green filter sets (supplied as standard) for Auramine, FITC, GFP and YFP (with blue filter set) plus Rhodamine, Texas Red and TRITC (with the green one), yet giving the possibility to combine any other specific filter sets for future upgrade.
Transmitted light through the exclusive X-LED⁸ to ensureMore Product Information