Mini automated cell counter, Moxi™ Z
Leverandør: Orflo
MXZ001
ORFLMXZ001EA 0 NOK
ORFLMXZ001
Mini automated cell counter, Moxi™ Z
Celletellere Elektroniske celletellere
The Moxi™ Z automated cell counter combines the the 'gold-standard' Coulter Principle typically used in high-end instruments with a patented thin-film sensor technology to provide highly accurate and repeatable direct cell/particle size and count over a broad dynamic range (2 to 34 µm), with single cell resolution.
Through the utilization of state of the art semiconductor, embedded processing and software algorithms the Moxi™ Z yields results in 8 to 15 seconds, with little to no sample prep required. The Moxi™ Z also provides a proprietary and revolutionary new insight into your cellMore Product Information