Human Tagged ORF Clone, Androgen Receptor (AR) (NM_000044)

Leverandør: OriGene

RC235415
ORIGRC235415EA 20600 NOK
ORIGRC235415
Human Tagged ORF Clone, Androgen Receptor (AR) (NM_000044)
Vektorer, plasmider og biblioteker

The ORF clone is ion-exchange column purified and shipped in a 2D barcoded Matrix tube containing 10 µg of transfection-ready, dried plasmid DNA (reconstitute with 100 µl of water).


  • Symbol: Androgen receptor
  • Synonyms: AIS; AR8; DHTR; HUMARA; HYSP1; KD; NR3C4; SBMA; SMAX1; TFM
  • E. coli selection: Kanamycin (25 µg/ml)
  • Mammalian cell selection: Neomycin
  • ORF size: 2760 bp
  • UniProt ID: P10275
  • Cytogenetics: Xq12
  • Domains: HOLI, Androgen_recep, zf-C4
  • Molecular weight: 99,2 kDa

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR