Human Recombinant Glycoprotein 2 (from HEK293T)
Leverandør: OriGene
TP307512
ORIGTP307512EA 14175 NOK
ORIGTP307512
Human Recombinant Glycoprotein 2 (from HEK293T)
Proteiner
This gene encodes an integral membrane protein that is secreted from intracellular zymogen granules and associates with the plasma membrane via glycosylphosphatidylinositol (GPI) linkage. The encoded protein binds pathogens such as enterobacteria, thereby playing an important role in the innate immune response. The C-terminus of this protein is related to the C-terminus of the protein encoded by the neighboring gene, uromodulin (UMOD). Alternative splicing results in multiple transcript variants.