Human Recombinant SNX18 (from HEK293T)

Leverandør: OriGene

TP319205
ORIGTP319205EA 14175 NOK
ORIGTP319205
Human Recombinant SNX18 (from HEK293T)
Proteiner

Purified recombinant protein of Homo sapiens sorting nexin 18 (SNX18), transcript variant 1.


  • Cytogenetics: 5q11.2
  • Buffer: 25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol


This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a SH3 domain. Multiple transcript variants encoding different isoforms have been found for this gene.


Preparation: Recombinant protein was captured through anti-DDK affinity column followed by conventional chromatography steps.
Stability: Stable for 12 months from the date of receipt of the product under proper storage and handling conditions. Avoid repeated freeze-thaw cycles.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR