Protective guards for conductivity cells and dissolved oxygen probes, Orion™
Leverandør: Thermo Orion
081045
ORIO081045EA 2925 NOK
ORIO081045
Protective guards for conductivity cells and dissolved oxygen probes, Orion™
Elektrodestativ
These protective guards provide additional protection and weight for conductivity cells and DO probes.
The plastic guard should be used when when minimal weight and protection are required. The combined guard with stainless steel cap and plastic-sleeve is ideal for use when weight is needed for submersion and standard protection is needed.
For use with:
Conductivity cells: 013005MD, 013010MD, 0130020MD, 013025MD, 013005A, 013010A, and 013005D
Dissolved oxygen probes: 080510MD, 0830005MD, 083010MD, 083025MD, 083060MD, 083005A, 083010A, and 083005D.