Soft-sided field case for portable meters, Orion™ Star series
Leverandør: Thermo Orion
1210005
ORIO1210005EA 1900 NOK
ORIO1210005
Soft-sided field case for portable meters, Orion™ Star series
Poser Equipment Bags and Instrument Cases
This cushioned field case with interior dividers and exterior pockets protects meters, electrodes, solution and beakers under field conditions.
For use with: Orion Star A120 series portable meters; Orion Star A220 series portable meters; Orion Star A320 series portable meters; Orion 3-Star, 4-Star and 5-Star series portable meters (retired); Orion Star and Star Plus portable meters.