No Cal stain lifter solution, Orion™
Leverandør: Thermo Orion
510015
ORIO510015EA 695 NOK
ORIO510015
No Cal stain lifter solution, Orion™
Rengjøringsløsninger Detergenter og overflatevasking
This cleaning solution restores proper electrode performance and prolongs the useful life of the electrode.