pH electrode, half-cell, Orion™ ROSS™ 8101BNWP
Leverandør: Thermo Orion
8101BNWP
ORIO8101BNWPEA 4350 NOK
ORIO8101BNWP
pH electrode, half-cell, Orion™ ROSS™ 8101BNWP
Elektroder pH-elektroder
This half-cell electrode provides stable readings to 0,01 pH in less than 30 seconds. Compared to conventional electrodes, the proven ROSS reference system exhibits superior stability in measurements, faster response, greater accuracy and precision when measuring samples that vary in temperature or when calibrating in temperatures that differ from the samples.
  • ROSS electrodes avoid the long term drift that other electrodes exhibit (ROSS electrodes drift by less than 0,002 pH per day), so recalibration is minimised
  • No sample contamination: ROSS electrodes