pH electrode, double junction, Orion™ 9102DJWP
Leverandør: Thermo Orion
9102DJWP
ORIO9102DJWPEA 3675 NOK
ORIO9102DJWP
pH electrode, double junction, Orion™ 9102DJWP
Elektroder pH-elektroder
This double junction pH electrode offers superior measurement stability, faster response, greater accuracy, longer life, least long-term drift and more reproducible results.
  • With waterproof BNC connector
  • Ag/AgCl double junction
  • Refillable