pH electrode, double junction, Orion™ 9110DJWP
Leverandør: Thermo Orion
9110DJWP
ORIO9110DJWPEA 5250 NOK
ORIO9110DJWP
pH electrode, double junction, Orion™ 9110DJWP
Elektroder pH-elektroder
This double junction pH electrode with semi-micro tip measures samples as small as 0,2 ml.
  • With waterproof BNC connector
  • Ag/AgCl double junction
  • Refillable