pH combination electrode with spear tip, Orion™ 91-63
Leverandør: Thermo Orion
9163SC
ORIO9163SCEA 5725 NOK
ORIO9163SC
pH combination electrode with spear tip, Orion™ 91-63
Elektroder pH-elektroder
This combination pH electrode with spear tip is ideal for piercing soft samples, such as cheese, meat, and fruit samples.
Reference system: Ag/AgCl