Ion selective electrode, half-cell, thiocyanate, Orion™
Leverandør: Thermo Orion
9458BN
ORIO9458BNEA 31475 NOK
ORIO9458BN
Ion selective electrode, half-cell, thiocyanate, Orion™
Elektroder ISE elektroder
This half-cell electrode is ideal for performing quick, accurate and economical measurements of thiocyanate ions in aqueous solutions.
  • Solid-state half-cell requires reference electrode
  • pH < 4 is recommended to avoid OH-interference