Orion™ Storage Solution for Ammonia Ion Selective Electrodes
Leverandør: Thermo Orion
951213
ORIO951213EA 1475 NOK
ORIO951213
Orion™ Storage Solution for Ammonia Ion Selective Electrodes
Elektrolytter Løsninger, lagring av elektroder
Take more accurate and reproducible ISE measurements with Thermo Scientific™ Orion™ ISE Ionic Strength Adjustors (ISA) and special reagents. Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes. Warranty: out of box.
  • Cat. No. 941709 is packaged as two 1 quart (2×1 L) bottles
  • Cat. No. 941609 is packaged as four 1 pint (4×475 ml) bottles
  • Cat. No. 940909 is packaged as one 1 gallon (3,7 L) bottle
  • Cat. No. 940999 is packaged as four 1 gallon (4×3,7 L) bottles
Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes with concentration 10 ppm.