Blood bank reagents
Leverandør: ORTHO CLINICAL DIAGNOSTICS
721105
ORTH721105EA 0 NOK
ORTH721105
Blood bank reagents
Diagnostiske systemer Clinical Reagents and Kits
Qualitative tests for analysing irregular blood group antibodies. Analysing and identifying irregular blood group antibodies are important procedures for ensuring optimum follow-up of transfusions, pregnancy and disease. Comparing the reactivity pattern of individual cells with the published ANTIGRAM® antigen profile helps with identifying antibodies.
  • Monoclonal antibodies for analysing various antigens on human erythrocytes