Native barcoding auxiliary kit v14, Oxford Nanopore Technologies
Leverandør: Oxford Nanopore Technologies
EXP-NBA114
OXNTEXP-NBA114EA 3575 NOK
OXNTEXP-NBA114
Native barcoding auxiliary kit v14, Oxford Nanopore Technologies
Nukleinsyre reagenser Next Generation Sequencing Reagents
This expansion is recommended for users who want to make use of unused barcodes to maximise the use of their Native Barcoding Kits.
This expansion pack contains additional library preparation reagents for the Native Barcoding Kit 24 and 96 V14 (SQK-NBD114.24 and SQK-NBD114.96) for another 12 reactions in conjunction with the Sequencing Auxiliary Vials V14 (EXP-AUX003).
The Native Barcoding Auxiliary Kit V14 (EXP-NBA114) can only be used with Kit 14 chemistry. This kit is not compatible with previous kits chemistries.
Bestillingsinformasjon: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
NB: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.