SFB expansion
Leverandør: Oxford Nanopore Technologies
EXP-SFB001
OXNTEXP-SFB001EA 370 NOK
OXNTEXP-SFB001
SFB expansion
Nukleinsyre reagenser Next Generation Sequencing Reagents
Short Fragment Buffer (SFB).
This kit is recommended for users who require additional Short Fragment Buffer for use with the 'PCR tiling of COVID-19 virus' protocol or any other experiment sequencing short fragments.
SFB Expansion contains four vials of Short Fragment Buffer (SFB), sufficient for four reactions.
Leveringsinformasjon: Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
NB: Oxford Nanopore Technologies products are currently for research use only.
Bestill nå
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.