Flow Cells for MinION™ and GridION™ Devices
Leverandør: Oxford Nanopore Technologies
FLO-MIN114 FLO-MIN114.8 FLOMIN004RAA.8 FLOMIN04RAA848 FLOMIN114.8.48 FLOMIN114.8.96 FLOMIN04RAA896 FLO-MIN004RAA
FLO-MIN114EA 8600 NOK
FLO-MIN114 FLO-MIN114.8 OXNTFLOMIN004RAA.8 OXNTFLOMIN04RAA848 OXNTFLOMIN114.8.48 OXNTFLOMIN114.8.96 OXNTFLOMIN04RAA896 FLO-MIN004RAA
Flow Cells for MinION™ and GridION™ Devices
Gjennomstrømmings celler
Nanopore sequencing flow cells for use with MinION™ and GridION™ devices.
Flow cells contain the proprietary sensor array, Application-Specific Integrated Circuit (ASIC), and nanopores for DNA/RNA sequencing on MinION™ and GridION™ devices.
R10.4.1 flow cells contain a double reader-head and are suitable for experiments where high consensus accuracy is required. Compatible with Kit 14 chemistry, R10.4.1 flow cells generate data at a modal accuracy above 99%. R9.4.1 flow cells are compatible with the previous generation of nanopore sequencing chemistries. These flow cells and associatedMore Product Information
Bestill nå
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.