Blood culture system, Signal®
Leverandør: THERMO FISHER DIAGNOSTICS
BC0100M BC0104A
OXOIBC0100MEA 3750 NOK
OXOIBC0100M OXOIBC0104A
Blood culture system, Signal®
Diagnostiske systemer Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Blood samples are collected from patients, using strict aseptic technique and sterile equipment. The samples are inoculated into the blood culture bottles and mixed with the medium.
  • Well-proven
  • Reliable
The formulation of the medium encourages the growth of aerobic, anaerobic and micro-aerophilic organisms. The medium is also designed to create pressure in the sealed bottle when organisms are growing. The detection of positive pressure is by means of a growth indicator device which is connected to the bottle after the blood sample is added.