Basisk Fuksin

Leverandør: THERMO FISHER DIAGNOSTICS
Danger

BR0050A
OXOIBR0050AEA 755 NOK
OXOIBR0050A
Basisk Fuksin
Basisk Fuksin
Formel: C₂₀H₂₀ClN₃
Smeltepunkt: 235 °C
Tetthet: 1,22 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012569
CAS-nummer: 632-99-5
EINECS: 211-189-6
Merck Indeks: 12,05686

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR