Antimicrobial susceptibility disks
Leverandør: THERMO FISHER DIAGNOSTICS
CT0002B
CT0003B
CT0005B
CT0007B
CT0010B
CT0011B
CT0013B
CT0015B
CT0016B
CT0017B
CT0018B
CT0019B
CT0020B
CT0021B
CT0023B
CT0024B
CT0025B
CT0026B
CT0028B
CT0030B
CT0031B
CT0032B
CT0033B
CT0034B
CT0036B
CT0037B
CT0038B
CT0040B
CT0041B
CT0042B
CT0043B
CT0044B
CT0046B
CT0047B
CT0048B
CT0051B
CT0052B
CT0054B
CT0056B
CT0057B
CT0058B
CT0059B
CT0060B
CT0061B
CT0063B
CT0064B
CT0067B
CT0069B
CT0072B
CT0076B
CT0088B
CT0091B
CT0096B
CT0104B
CT0107B
CT0119B
CT0123B
CT0124B
CT0127B
CT0149B
CT0152B
CT0159B
CT0161B
CT0166B
CT0167B
CT0180B
CT0183B
CT0188B
CT0193B
CT0199B
CT0207B
CT0223B
CT0225B
CT0232B
CT0249B
CT0263B
CT0264B
CT0407B
CT0411B
CT0412B
CT0417B
CT0424B
CT0425B
CT0434B
CT0446B
CT0453B
CT0455B
CT0466B
CT0493B
CT0520B
CT0523B
CT0538B
CT0545B
CT0623B
CT0639B
CT0647B
CT0653B
CT0661B
CT0664B
CT0665B
CT0666B
CT0668B
CT0693B
CT0725B
CT0758B
CT0774B
CT0794B
CT0998B
CT0771B
CT0906B
CT1587B
CT1612B
CT1629B
CT1633B
CT1650B
CT1662B
CT1751B
CT1754B
CT1756B
CT1761B
CT1806B
CT1807B
CT0823B
CT1616B
CT1841B
CT1880B
CT1897B
CT1758B
OXOICT0002BEA
470
NOK
OXOICT0002B
OXOICT0003B
OXOICT0005B
OXOICT0007B
OXOICT0010B
OXOICT0011B
OXOICT0013B
OXOICT0015B
OXOICT0016B
OXOICT0017B
OXOICT0018B
OXOICT0019B
OXOICT0020B
OXOICT0021B
OXOICT0023B
OXOICT0024B
OXOICT0025B
OXOICT0026B
OXOICT0028B
OXOICT0030B
OXOICT0031B
OXOICT0032B
OXOICT0033B
OXOICT0034B
OXOICT0036B
OXOICT0037B
OXOICT0038B
OXOICT0040B
OXOICT0041B
OXOICT0042B
OXOICT0043B
OXOICT0044B
OXOICT0046B
OXOICT0047B
OXOICT0048B
OXOICT0051B
OXOICT0052B
OXOICT0054B
OXOICT0056B
OXOICT0057B
OXOICT0058B
OXOICT0059B
OXOICT0060B
OXOICT0061B
OXOICT0063B
OXOICT0064B
OXOICT0067B
OXOICT0069B
OXOICT0072B
OXOICT0076B
OXOICT0088B
OXOICT0091B
OXOICT0096B
OXOICT0104B
OXOICT0107B
OXOICT0119B
OXOICT0123B
OXOICT0124B
OXOICT0127B
OXOICT0149B
OXOICT0152B
OXOICT0159B
OXOICT0161B
OXOICT0166B
OXOICT0167B
OXOICT0180B
OXOICT0183B
OXOICT0188B
OXOICT0193B
OXOICT0199B
OXOICT0207B
OXOICT0223B
OXOICT0225B
OXOICT0232B
OXOICT0249B
OXOICT0263B
OXOICT0264B
OXOICT0407B
OXOICT0411B
OXOICT0412B
OXOICT0417B
OXOICT0424B
OXOICT0425B
OXOICT0434B
OXOICT0446B
OXOICT0453B
OXOICT0455B
OXOICT0466B
OXOICT0493B
OXOICT0520B
OXOICT0523B
OXOICT0538B
OXOICT0545B
OXOICT0623B
OXOICT0639B
OXOICT0647B
OXOICT0653B
OXOICT0661B
OXOICT0664B
OXOICT0665B
OXOICT0666B
OXOICT0668B
OXOICT0693B
OXOICT0725B
OXOICT0758B
OXOICT0774B
OXOICT0794B
OXOICT0998B
OXOICT0771B
OXOICT0906B
OXOICT1587B
OXOICT1612B
OXOICT1629B
OXOICT1633B
OXOICT1650B
OXOICT1662B
OXOICT1751B
OXOICT1754B
OXOICT1756B
OXOICT1761B
OXOICT1806B
OXOICT1807B
OXOICT0823B
OXOICT1616B
OXOICT1841B
OXOICT1880B
OXOICT1897B
OXOICT1758B
Antimicrobial susceptibility disks
Mikrobiologiske testsystemer
Antibiotiske tester
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use antimicrobial susceptibility disks (5×50).
- Reagent code and concentration printed clearly on each disk
- Shatterproof, spring-loaded cartridges for easy dispensing
- Cartridges individually blister-packed with desiccant to maintain product quality
SPESIFIKASJONER
Environmentally Preferable |