Oxoid biochemical identification system (O.B.I.S.)
Leverandør: THERMO FISHER DIAGNOSTICS
ID0570M ID0580M ID0600M
OXOIID0570MEA 3175 NOK
OXOIID0570M OXOIID0580M OXOIID0600M
Oxoid biochemical identification system (O.B.I.S.)
Mikrobiologiske testsystemer Enzymatiske tester
The Oxoid Biochemical Identification System (O.B.I.S.) is a rapid colorimetric test for the determination of biochemical activity in microorganisms. It is a rapid screening method for differentiation of other species.
  • Reduces the need for full biochemical identification of all suspect colonies