Natriumklorid

Supplier: THERMO FISHER DIAGNOSTICS

LP0005B
OXOILP0005BEA 515 NOK
OXOILP0005B
Natriumklorid
Natriumklorid
Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2,16 g/cm³ (20 °C)
Storage Temperature: Romtemperatur
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR