Laktose monohydrat

Leverandør: THERMO FISHER DIAGNOSTICS

LP0070B
OXOILP0070BEA 575 NOK
OXOILP0070B
Laktose monohydrat
Laktose monohydrat
Formel: C₁₂H₂₂O₁₁·1H₂O
Smeltepunkt: 223 °C
Lagringstemperatur: Romtemperatur
CAS-nummer: 10039-26-6
EINECS: 200-559-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR