Natriumhydrogenselenitt

Leverandør: THERMO FISHER DIAGNOSTICS
Danger

LP0121A
OXOILP0121AEA 1650 NOK
OXOILP0121A
Natriumhydrogenselenitt
Natriumhydrogenselenitt
Formel: HNaO₃Se
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003525
CAS-nummer: 7782-82-3
EINECS: 231-966-3
FN: 2630
ADR: 6.1,I

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR