Microbact™
Leverandør: THERMO FISHER DIAGNOSTICS
MB1131A
OXOIMB1131AEA 4425 NOK
OXOIMB1131A
Microbact™
Mikrobiologiske testsystemer Enzymatiske tester
The Microbact™ Gram-negative system is to be used for the identification of aerobic and facultatively anaerobic Gram-negative bacteria (Enterobacteriaceae and miscellaneous Gram- negative bacteria). Organism identification is based on pH change and substrate utilisations as established by published reference methodologies.
  • Fast
  • Simple
  • Reliable