Bromocresol Purple (BCP) Agar
Leverandør: THERMO FISHER DIAGNOSTICS
PO5118A
OXOIPO5118AEA 0 NOK
OXOIPO5118A
Bromocresol Purple (BCP) Agar
Mikrobiologiske medier Media, preparerte for mikrobiologi
A medium for the isolation and differentiation of lactose positive and lactose negative enterobacteriaceae.