Latex agglutination test, Wellcolex®
Leverandør: THERMO FISHER DIAGNOSTICS
R30858302 R30858401 R30368701 R30858301
OXOIR30858302EA 11525 NOK
OXOIR30858302 OXOIR30858401 OXOIR30368701 OXOIR30858301
Latex agglutination test, Wellcolex®
Mikrobiologiske testsystemer Antigen-antistoff tester Ag-AK, Latex
Wellcolex® Rapid Latex Agglutination test kits are designed for rapid and accurate identification of enteric bacteria, such as Salmonella, Shigella and E.coli. With its high positive and negative predictive values (PPV >98,7%) and results in less than three minutes, laboratories can use this rapid screening test to eliminate Salmonella-negative samples from further testing, reducing the number of samples requiring confirmatory testing.
  • Easy to use
  • Save up to two days' testing time over traditional screening procedures