Culti-Loops™ Bacteroides fragilis ATCC® 25285™
Leverandør: THERMO FISHER DIAGNOSTICS
R4601250
OXOIR4601250EA 3225 NOK
OXOIR4601250
Culti-Loops™ Bacteroides fragilis ATCC® 25285™
Mikrobiologiske testsystemer Tester for mikroorganismer
Culti-Loops™ QC organisms. Bacteroides fragilis ATCC® 25285™ is recommended to be used for culture media and antimicrobial susceptibility quality control testing.
Bacteroides fragilis ATCC® 25285™ is recommended to be used for culture media and antimicrobial susceptibility quality control testing by CLSI. β-lactamase positive strain. The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Forpakning: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.