Culti-Loops™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™
Leverandør: THERMO FISHER DIAGNOSTICS
R4606000
OXOIR4606000EA 3225 NOK
OXOIR4606000
Culti-Loops™ Salmonella enterica subsp. enterica serovar Typhimurium ATCC™ 14028™
Mikrobiologiske testsystemer Tester for mikroorganismer
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Forpakning: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.