Microorganisms strains, Culti-Loops®
Leverandør: THERMO FISHER DIAGNOSTICS
R4607011
OXOIR4607011EA 3225 NOK
OXOIR4607011
Microorganisms strains, Culti-Loops®
Mikrobiologiske testsystemer Tester for mikroorganismer
Culti-Loops® are ready-to-use, disposable inoculation loops containing stabilized, preserved, viable microorganisms. A quality source of stock organisms, Culti-Loops® are derived from American Type Culture Collection (ATCC®) or other referenced sources. Culti-Loops® are manufactured in an ISO-certified facility using our proprietary preservation technique, ensuring enhanced recovery and reproducibility. The loop may be dissolved in rehydration fluid or streaked directly onto appropriate media.
  • Convenient, ready-to-use, disposable loops
  • Organisms include bacterial, fungal, and mycobacterium isolates
  • Individually wrapped in foil-sealed pouches to preserve viability
  • Peel-off label for convenient documentation
  • No glass, needles, or forceps ensures user safety