Culti-Loops™ Neisseria meningitidis serogp B ATCC™ 13090™
Leverandør: THERMO FISHER DIAGNOSTICS
R4607049
OXOIR4607049EA 3225 NOK
OXOIR4607049
Culti-Loops™ Neisseria meningitidis serogp B ATCC™ 13090™
Mikrobiologiske testsystemer Tester for mikroorganismer
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Forpakning: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.