Culti-Loops™ Peptostreptococcus anaerobius ATCC™ 27337™
Leverandør: THERMO FISHER DIAGNOSTICS
R4607051
OXOIR4607051EA 3225 NOK
OXOIR4607051
Culti-Loops™ Peptostreptococcus anaerobius ATCC™ 27337™
Mikrobiologiske testsystemer Tester for mikroorganismer
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium.
Forpakning: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.